KEY465196393 Key Organics / BIONET

4-(4-heptylphenyl)-2-methyl-5-oxo-5H-indeno[1,2-b]pyridine-3-carbonitrile

  • MDL number MFCD03012571
  • Cas Number : 439108-64-2
  • Molecular Weight : 394.518

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/76844

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4T-0296_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart