KEY465196398 Key Organics / BIONET

1-[(2,6-dichlorophenyl)methyl]-1H-imidazole-4,5-dicarbonitrile

  • MDL number MFCD03012702
  • Cas Number : 439108-73-3
  • Molecular Weight : 277.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/76864

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4T-0319_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart