KEY465196404 Key Organics / BIONET

5-(2H-1,3-benzodioxol-5-yl)-1-(4-chlorobenzenesulfonyl)-1H-pyrazole

  • MDL number MFCD03617362
  • Cas Number : 956193-64-9
  • Molecular Weight : 362.78

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/76887

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4T-0352_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart