KEY465196409 Key Organics / BIONET

2-(4-nitrophenyl)imidazo[1,2-a]pyrimidine

  • MDL number MFCD00602988
  • Cas Number : 28266-96-8
  • Molecular Weight : 240.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/76898

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4T-0608_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart