KEY465196431 Key Organics / BIONET

4-ethyl-5-{2-[3-methyl-5-(trifluoromethyl)-1H-pyrazol-1-yl]-1,3-thiazol-4-yl}-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD03012721
  • Cas Number : 956193-65-0
  • Molecular Weight : 360.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/76964

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4T-0845_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart