KEY465196436 Key Organics / BIONET

5,7-dimethyl-6-[(2E)-3-phenylprop-2-en-1-yl]-[1,2,4]triazolo[1,5-a]pyrimidin-2-amine

  • MDL number MFCD03012729
  • Cas Number : 691887-98-6
  • Molecular Weight : 279.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/76990

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4T-0886_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart