KEY465196458 Key Organics / BIONET

2-{[(5Z)-4-oxo-5-{[2-(1H-pyrazol-1-yl)phenyl]methylidene}-4,5-dihydro-1,3-thiazol-2-yl]amino}propanoic acid

  • MDL number MFCD03848543
  • Molecular Weight : 342.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/77107

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4W-0301_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart