KEY465196466 Key Organics / BIONET

2-{3-oxo-2H,3H,4H-pyrido[3,2-b][1,4]oxazin-2-yl}ethyl 4-fluorobenzoate

  • MDL number MFCD03787429
  • Cas Number : 866018-89-5
  • Molecular Weight : 316.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/77130

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4W-0338_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart