KEY465196487 Key Organics / BIONET

1,3-diethyl 2-[6-chloro-4-(trifluoromethyl)pyridin-2-yl]-2-ethylpropanedioate

  • MDL number MFCD08281856
  • Cas Number : 882747-55-9
  • Molecular Weight : 367.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/77177

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4W-0830_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart