KEY465196496 Key Organics / BIONET

3-(2-fluorophenyl)-3-[2-(2,2,2-trifluoroacetyl)-1H-pyrrol-1-yl]propanoic acid

  • MDL number MFCD03848564
  • Cas Number : 866019-25-2
  • Molecular Weight : 329.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/77195

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4W-0851_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart