KEY465196516 Key Organics / BIONET

3-hydroxy-6-(hydroxymethyl)-2-[(4-methylpiperidin-1-yl)methyl]-4H-pyran-4-one

  • MDL number MFCD02081501
  • Cas Number : 92198-59-9
  • Molecular Weight : 253.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/77238

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/4X-0209_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart