KEY465196517 Key Organics / BIONET

5-[3-(trifluoromethyl)phenyl]pyrazolo[1,5-a]pyrimidine-3-carbonitrile

  • MDL number MFCD04125471
  • Cas Number : 866019-49-0
  • Molecular Weight : 288.233

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/77240

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4X-0215_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart