KEY465196523 Key Organics / BIONET

2-(5-oxo-3-phenyl-2,5-dihydro-1H-pyrazol-1-yl)-6-phenyl-3,4-dihydropyrimidin-4-one

  • MDL number MFCD04125484
  • Cas Number : 866019-63-8
  • Molecular Weight : 330.347

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/77255

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4X-0245_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart