KEY465196527 Key Organics / BIONET

(2E)-3-{[5-(carboxymethyl)-2-hydroxyphenyl]carbamoyl}prop-2-enoic acid

  • MDL number MFCD04125489
  • Cas Number : 866019-68-3
  • Molecular Weight : 265.221

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/77260

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4X-0258_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart