KEY465196536 Key Organics / BIONET

(2Z)-2-{4-[2-(1,3-dioxo-2,3-dihydro-1H-isoindol-2-yl)ethyl]-1,3-thiazol-2-yl}-3-(4-methylphenyl)prop-2-enenitrile

  • MDL number MFCD04125504
  • Cas Number : 1164562-29-1
  • Molecular Weight : 399.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/77277

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4X-0298_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart