KEY465196539 Key Organics / BIONET

(2E)-3-[2-(2,4-dichlorophenoxy)phenyl]-2-(thiophen-2-yl)prop-2-enenitrile

  • MDL number MFCD04125513
  • Cas Number : 866019-88-7
  • Molecular Weight : 372.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/77286

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4X-0313_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart