KEY465196542 Key Organics / BIONET

N'-[(1Z)-[2-(2,4-dichlorophenoxy)phenyl]methylidene]-1-methyl-1H-pyrrole-2-carbohydrazide

  • MDL number MFCD05975068
  • Cas Number : 866019-92-3
  • Molecular Weight : 388.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/77290

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4X-0318_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart