KEY465196566 Key Organics / BIONET

methyl 2-amino-4-(piperidin-1-yl)thieno[2,3-d]pyrimidine-6-carboxylate

  • MDL number MFCD04125546
  • Cas Number : 866020-42-0
  • Molecular Weight : 292.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/77347

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4X-0833_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart