KEY465196577 Key Organics / BIONET

(2E)-2-[(E)-benzoyl]-3-[4-(2-chloroacetyl)-1H-pyrrol-2-yl]prop-2-enenitrile

  • MDL number MFCD04125566
  • Cas Number : 866038-40-6
  • Molecular Weight : 298.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/77376

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4X-0875_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart