KEY465196580 Key Organics / BIONET

(3Z)-3-[(4-hydroxy-3-nitrophenyl)methylidene]-2,3-dihydro-1H-indol-2-one

  • MDL number MFCD05668780
  • Cas Number : 866038-46-2
  • Molecular Weight : 282.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/77385

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4X-0886_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart