KEY465196589 Key Organics / BIONET

5-phenyl-2-(pyridin-3-yl)-1,3-thiazol-4-ol

  • MDL number MFCD00895727
  • Cas Number : 131786-81-7
  • Molecular Weight : 254.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/77435

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4Y-0813_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart