KEY465196593 Key Organics / BIONET

(3Z)-6-chloro-3-[(4-chloro-3-nitrophenyl)methylidene]-3,4-dihydro-2H-1-benzothiopyran-4-one

  • MDL number MFCD07021460
  • Cas Number : 900019-21-8
  • Molecular Weight : 366.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/77449

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4Y-0923_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart