KEY465196600 Key Organics / BIONET

methyl 3-[4-(2,2-dimethylpropanamido)phenoxy]thiophene-2-carboxylate

  • MDL number MFCD07021469
  • Cas Number : 900019-28-5
  • Molecular Weight : 333.40

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/77461

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4Y-5024_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart