KEY465196632 Key Organics / BIONET

1-(4-chlorophenyl)-3-[(3,4-dichloro-1-methyl-1H-pyrazol-5-yl)(methyl)amino]urea

  • MDL number MFCD00127508
  • Cas Number : 318284-27-4
  • Molecular Weight : 348.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/77573

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5B-076_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart