KEY465196636 Key Organics / BIONET

3-(phenylamino)-3H,4H-thieno[3,2-d]pyrimidin-4-one

  • MDL number MFCD00138936
  • Cas Number : 338750-82-6
  • Molecular Weight : 243.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/77586

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5B-095_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart