KEY465196642 Key Organics / BIONET

(4Z)-1-(4-chlorophenyl)-3-ethoxy-4-{[(morpholin-4-yl)amino]methylidene}-4,5-dihydro-1H-pyrazol-5-one

  • MDL number MFCD00138439
  • Cas Number : 338751-09-0
  • Molecular Weight : 350.80

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/77632

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5C-081_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart