KEY465196662 Key Organics / BIONET

6-(4-chlorophenyl)-2-{[(2E)-3-phenylprop-2-en-1-yl]sulfanyl}-[1,2,4]triazolo[1,5-a]pyrimidin-7-amine

  • MDL number MFCD00172152
  • Cas Number : 338751-64-7
  • Molecular Weight : 393.89

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/77695

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5D-031_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart