KEY465196666 Key Organics / BIONET

methyl 1-(2,4-dichlorophenyl)-4-hydroxy-6-oxo-1,6-dihydropyridazine-3-carboxylate

  • MDL number MFCD00141007
  • Cas Number : 121582-57-8
  • Molecular Weight : 315.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/77712

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5D-058_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart