KEY465196668 Key Organics / BIONET

3-(2-chlorophenoxy)-6-phenylpyridazine-4-carbonitrile

  • MDL number MFCD00794592
  • Cas Number : 338751-75-0
  • Molecular Weight : 307.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/77720

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5D-074_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart