KEY465196670 Key Organics / BIONET

3-(4-methylphenoxy)-6-phenylpyridazine-4-carbonitrile

  • MDL number MFCD00974820
  • Cas Number : 338752-71-9
  • Molecular Weight : 287.322

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/77722

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5D-077_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart