KEY465196689 Key Organics / BIONET

4-(2,3-dichlorobenzoyl)-1H-pyrrole-2-carboxamide

  • MDL number MFCD00141907
  • Cas Number : 338753-53-0
  • Molecular Weight : 283.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/77784

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5D-175_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart