KEY465196730 Key Organics / BIONET

6'-(4-chlorophenyl)-2,4-dihydrospiro[1-benzopyran-3,3'-[1,5]dioxane]-4-one

  • MDL number MFCD00139869
  • Cas Number : 338755-68-3
  • Molecular Weight : 330.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/77962

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5E-914_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart