KEY465196733 Key Organics / BIONET

methyl (3E)-3-(methoxyimino)-2-(phenylformamido)propanoate

  • MDL number MFCD00141012
  • Cas Number : 318284-57-0
  • Molecular Weight : 250.254

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/77969

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5E-923_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart