KEY465196735 Key Organics / BIONET

3-methyl-1-(4-methylphenyl)-4,5-dihydro-1H-pyrazol-5-one

  • MDL number MFCD00035708
  • Cas Number : 86-92-0
  • Molecular Weight : 188.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-methyl-2-(4-methylphenyl)-1,2-dihydro-3H-pyrazol-3-one
Inchi Code 1S/C11H12N2O/c1-8-3-5-10(6-4-8)13-11(14)7-9(2)12-13/h3-7,12H,1-2H3
Inchi Key YAVQZXBVUORYNQ-UHFFFAOYSA-N

Description

Purity 90%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/77974

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/5E-930_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart