KEY465196753 Key Organics / BIONET

2-(2,6-difluorophenyl)-5-methyl-1,3-thiazol-4-ol

  • MDL number MFCD00098886
  • Cas Number : 271772-57-7
  • Molecular Weight : 227.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/78012

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5F-040_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart