KEY465196757 Key Organics / BIONET

5-[(E)-2-[5-(2,4-dichlorophenyl)furan-2-yl]ethenyl]-3-methyl-4-nitro-1,2-oxazole

  • MDL number MFCD00214763
  • Cas Number : 338756-56-2
  • Molecular Weight : 365.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/78018

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5F-047_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart