KEY465196761 Key Organics / BIONET

methyl 3-[(4-methoxyphenyl)methoxy]thieno[2,3-b]pyridine-2-carboxylate

  • MDL number MFCD00141083
  • Cas Number : 338756-97-1
  • Molecular Weight : 329.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/78042

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5F-325S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart