KEY465196762 Key Organics / BIONET

methyl 3-[(1-ethoxy-1-oxopropan-2-yl)oxy]thieno[2,3-b]pyridine-2-carboxylate

  • MDL number MFCD01936301
  • Cas Number : 400083-29-6
  • Molecular Weight : 309.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/78045

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/5F-328S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart