KEY465196773 Key Organics / BIONET

methyl 5-bromo-2-oxo-1-{[3-(trifluoromethyl)phenyl]methyl}-1,2-dihydropyridine-3-carboxylate

  • MDL number MFCD01936302
  • Cas Number : 400083-32-1
  • Molecular Weight : 390.156

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/78071

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5F-365S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart