KEY465196779 Key Organics / BIONET

1-methyl-5-(3-nitrophenoxy)-3-(trifluoromethyl)-1H-pyrazole-4-carbonitrile

  • MDL number MFCD00141028
  • Cas Number : 318469-12-4
  • Molecular Weight : 312.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/78087

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/5F-384S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart