KEY465196802 Key Organics / BIONET

1-[2,6-dinitro-4-(trifluoromethyl)phenyl]piperidin-4-one

  • MDL number MFCD00231571
  • Cas Number : 338757-95-2
  • Molecular Weight : 333.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/78156

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/5F-946_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart