KEY465196853 Key Organics / BIONET

N-phenyl-2-(1,2,3,4-tetrahydroisoquinolin-1-yl)acetamide

  • MDL number MFCD00215028
  • Cas Number : 338761-98-1
  • Molecular Weight : 266.344

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/78419

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5H-016_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart