KEY465196860 Key Organics / BIONET

(2Z)-3-{[3-chloro-5-(trifluoromethyl)pyridin-2-yl]sulfanyl}-1-(4-chlorophenyl)prop-2-en-1-one

  • MDL number MFCD00244261
  • Cas Number : 288270-43-9
  • Molecular Weight : 378.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/78441

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5H-045_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart