KEY465196866 Key Organics / BIONET

3-chloro-2-(pyridine-3-carbonyl)-5-(trifluoromethyl)pyridine

  • MDL number MFCD00244682
  • Cas Number : 338953-55-2
  • Molecular Weight : 286.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/78466

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/5H-076_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart