KEY465196885 Key Organics / BIONET

1-(4,6-dimethylpyridin-2-yl)-4,5-dihydroxy-3-methylimidazolidin-2-one

  • MDL number MFCD04125585
  • Cas Number : 866038-53-1
  • Molecular Weight : 237.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/78540

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5H-423S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart