KEY465196890 Key Organics / BIONET

4-chloro-N-{1-[4-ethyl-5-(methylsulfanyl)-4H-1,2,4-triazol-3-yl]ethyl}benzene-1-sulfonamide

  • MDL number MFCD00202159
  • Cas Number : 338954-07-7
  • Molecular Weight : 360.88

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/78553

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5H-436S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart