KEY465196902 Key Organics / BIONET

3-chloro-N-[2-methyl-2-(methylsulfanyl)propyl]-5-(trifluoromethyl)pyridin-2-amine

  • MDL number MFCD00975572
  • Cas Number : 338954-37-3
  • Molecular Weight : 298.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/78595

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5J-009_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart