KEY465196903 Key Organics / BIONET

(2E)-3-(dimethylamino)-2-{5-[(1Z)-1-(methoxyimino)ethyl]thiophen-2-yl}prop-2-enenitrile

  • MDL number MFCD00665062
  • Cas Number : 338954-44-2
  • Molecular Weight : 249.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/78602

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5J-018_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart