KEY465196921 Key Organics / BIONET

1-[(3-methoxyphenyl)methyl]-5,6-dimethyl-4-nitro-1H-1,3-benzodiazole

  • MDL number MFCD00231803
  • Cas Number : 338954-64-6
  • Molecular Weight : 311.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/78656

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/5J-316S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart