KEY465196934 Key Organics / BIONET

1-[(4-methoxyphenyl)methyl]-2,5,6-trimethyl-4-nitro-1H-1,3-benzodiazole

  • MDL number MFCD00232102
  • Cas Number : 338955-07-0
  • Molecular Weight : 325.368

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/78711

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5J-380S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart